Learn More
Cayman Chemical ANTIBODY RIbavIrIn 500mg
SDP
Supplier:  Cayman Chemical 16757500 MG
Encompass_Preferred
An antiviral nucleoside analog; metabolized to an active triphosphate form that induces viral chain termination and inhibits viral polymerases; reduces replication in a panel of seven RNA and four DNA viruses in Vero cells (EC50s = 2-95 UG/ml); reduces replication of SARS-CoV-2 in Vero cells (EC50 = 109.5 µM); aerosol administration reduces mortality in a mouse model of influenza A infection at 30 mg/kg
Catalog No. 50-297-9606