Learn More
Cayman Chemical ANTIBODY RIbocIl 10mg
SDP
Supplier:  Cayman Chemical 1848710 MG
Encompass_Preferred
An inhibitor of FMN riboswitch-mediated expression of ribB, a gene responsible for the synthesis and transport of riboflavin; depletes cellular levels of riboflavin (IC50 = 0.3 µM) and inhibits E. coli growth (MIC = 2 UG/ml)
Catalog No. 50-297-9373