Learn More
Cayman Chemical ANTIBODY RIfabutIn 10mg
SDP
Supplier:  Cayman Chemical 1646810 MG
Encompass_Preferred
A rifamycin antibiotic; active against 81 clinical isolates of H. pylori (MIC50 = 0.25 UG/ml), as well as S. aureus, S. pyogenes, and C. trachomatis (MICs = 0.004, 0.005, and ~0.008 UG/ml, respectively); active against 302 strains of M. tuberculosis at 0.5 UG/ml; inhibits the activity of RNAP; reduces the number of spleen, lung, and liver CFUs in mouse models of disseminated MAC infection
Catalog No. 50-296-9862