Learn More
Cayman Chemical ANTIBODY RIvenprost 100ug
SDP
Supplier:  Cayman Chemical 13618100 UG
Encompass_Preferred
An EP4 receptor agonist (Kis = 0.7, 56, 620, and >10,000 nM, respectively, as the free acid); enhances bone morphogenic protein-induced increases in alkaline phosphatase activity in primary mouse calvarial osteoblasts at 100 nM; decreases LPS-induced increases in plasma TNF-α levels in rats; decreases trabecular separation, as well as increases bone volume, in rats at 10 UG/kg; increases proximal tibiae biomechanical strength in ovariectomized rats at 3 and 10 mg/kg
Catalog No. 50-295-5394