Learn More
Cayman Chemical ANTIBODY RM 133-3 10mg
SDP
Supplier:  Cayman Chemical 3891710 MG
Encompass_Preferred
An ionizable cationic lipid; has been used in the formation of LNPs for the delivery of mRNA in vivo; LNPs containing RM 133-3 and encapsulating human EPO mRNA increase serum human EPO protein levels in mice
Catalog No. 50-291-8819