Learn More
Cayman Chemical ANTIBODY RN-1747 5mg
SDP
Supplier:  Cayman Chemical 375735 MG
Encompass_Preferred
A TRPV4 agonist; selectively induces calcium influx in HEK293 cells expressing human, rat, or mouse TRPV4 (EC50s = 0.77, 4.1, and 4 µM, respectively) over HEK293 cells expressing human TRPV1, TRPV3, or TRPM8 (EC50s = >100, >30, and >30 µM, respectively); induces inward current in Xenopus oocytes expressing human TRPV4 at 10 µM; topical administration of RN-1747 induces hypothermia in rats
Catalog No. 50-291-7719