Learn More
Cayman Chemical ANTIBODY RN-18 25mg
SDP
Supplier:  Cayman Chemical 2333125 MG
Encompass_Preferred
An HIV-1 Vif inhibitor; inhibits Vif-mediated degradation of the DNA editing enzyme APOBEC3G (IC50s = 10-30 µM); inhibits HIV-1 viral replication (IC50s = 4.5 and 10 µM for nonpermissive CEM and H9 cells, respectively, and >100 µM for permissive cells)
Catalog No. 50-299-3816