Learn More
Cayman Chemical ANTIBODY Ro 4929097 5mg
SDP
Supplier:  Cayman Chemical 199965 MG
Encompass_Preferred
A potent, selective inhibitor of γ-secretase (IC50 = 4 nM) that blocks both Aβ40 and Notch processing (IC50s = 14 and 5 nM, respectively); orally active against cancer xenograft models, altering gene expression related to angiogenesis and cell differentiation
Catalog No. 50-298-4568