Learn More
Cayman Chemical ANTIBODY RO4987655 25mg
SDP
Supplier:  Cayman Chemical 1948825 MG
Encompass_Preferred
A MEK inhibitor (IC50 = 5 nM); selective for MEK over 400 kinases at 10 µM; inhibits proliferation of COLO 205, HT-29, QG56, MIA PaCa-2, and C32 cells (IC50s = 0.86, 1.7, 9.5, 3.3, and 8.4 nM, respectively); reduces tumor growth in a variety of mouse xenograft models; inhibits the phosphorylation of ERK in tumor tissue in an HT-29 mouse xenograft model at 6.25 mg/kg per day; acts synergistically with everolimus to reduce tumor volume in an HCT116 mouse xenograft model
Catalog No. 50-298-0686