Learn More
Cayman Chemical ANTIBODY RoquInImex 100mg
SDP
Supplier:  Cayman Chemical 30206100 MG
Encompass_Preferred
An immunomodulator with diverse biological activities; increases the number of myocardial MHC class II-expressing cells, the ratio of cytotoxic to helper T cells, and survival and decreases necrotic lesion area in a mouse model of murine coxsackievirus B3-induced myocarditis at 80 mg/kg; induces IFN-γ production, reduces TNF-α and IL-1β production, prevents the development of proteinuria, and ameliorates nephritis in a mouse model of chronic GVHD; decreases colonic MPO activity and mucosal damage in a mouse model of DSS-induced colitis at 300 mg/kg; decreases tumor weight and tumor blood flow in a VHL type 2a paraganglioma mouse xenograft model
Catalog No. 50-298-8442