Learn More
Cayman Chemical ANTIBODY RS 504393 1mg
SDP
Supplier:  Cayman Chemical 173301 MG
Encompass_Preferred
A selective antagonist of CCR2 (IC50s = 89 nM); inhibits MCP-1-stimulated chemotaxis (IC50 = 330 nM) and ischemia-reperfusion injury in kidneys
Catalog No. 50-298-0178