Learn More
Cayman Chemical ANTIBODY RSC-133 10mg
SDP
Supplier:  Cayman Chemical 900183910 MG
Encompass_Preferred
Promotes the reprogramming of human somatic cells to pluripotent stem cells; increases the number of human foreskin fibroblasts that express alkaline phosphatase when used at 10 μM with four standard reprogramming factors; down-regulates inducers of cellular senescence and inhibits Dnmt1 and HDAC1
Catalog No. 50-289-5070