Learn More
Cayman Chemical ANTIBODY RX-3117 1mg
SDP
Supplier:  Cayman Chemical 279221 MG
Encompass_Preferred
A nucleoside derivative and an anticancer agent; cytotoxic to HCT116, MDA-MB-231, PANC-1, A549, and U251 cancer cells (IC50s = 0.39, 0.18, 0.62, 0.34, and 0.83 µM, respectively); induces DNA damage and apoptosis in A2780 cells at 1 and 10 µM; reduces mean tumor volume in HCT116, HT-29, Caki-1, MIA PaCa-2, and BxPC-3 mouse xenograft models from 50-500 mg/kg
Catalog No. 50-293-7267