Learn More
Cayman Chemical ANTIBODY RY785 5mg
SDP
Supplier:  Cayman Chemical 198135 MG
Encompass_Preferred
A selective inhibitor of Kv2.1 voltage-gated potassium channels (IC50 = 0.05 µM)
Catalog No. 50-298-1893