Learn More
Cayman Chemical ANTIBODY Saclofen 5mg
SDP
Supplier:  Cayman Chemical 173555 MG
Encompass_Preferred
A sulfonic analog of GABA that acts as a competitive antagonist of the GABAB receptor (IC50 = 7.8 µM)
Catalog No. 50-296-7328