Learn More
Cayman Chemical ANTIBODY Safngol 25mg
SDP
Supplier:  Cayman Chemical 1862425 MG
Encompass_Preferred
A dual inhibitor of PKC and SPHK; inhibits PKC by binding at the regulatory phorbol-binding domain (IC50 = 24 µM); inhibits SPHK (Ki = ~5 µM); restores sensitivity to cisplatin in resistant AGScis5 cells and N87 gastric cancer cells from 0.2-6 µM
Catalog No. 50-296-6002