Learn More
Cayman Chemical ANTIBODY SAHA-d5 500ug
SDP
Supplier:  Cayman Chemical 22107500 UG
Encompass_Preferred
An internal standard for the quantification of SAHA by GC- or LC-MS
Catalog No. 50-298-6200