Learn More
Cayman Chemical ANTIBODY SAINT C-18 1mg
SDP
Supplier:  Cayman Chemical 397091 MG
Encompass_Preferred
A cationic lipid; has been used in the formation of liposomes for the delivery of plasmid DNA, proteins, siRNA, and small molecules; complexes of SAINT C-18 with plasmid DNA encoding PEDF inhibit corneal neovascularization induced by complexes of SAINT C-18 with plasmid DNA encoding FGF2 in rats
Catalog No. 50-291-9257