Learn More
Cayman Chemical ANTIBODY SalermIde 50mg
SDP
Supplier:  Cayman Chemical 1317850 MG
Encompass_Preferred
An inhibitor of SIRT1 and SIRT2, causing tumor-specific apopoptic cell death; causes 90% apoptosis within 72 hours (IC50 ~20 μM) by reactivating proapototic genes that are repressed by SIRT1 in MOLT4 leukemia cells
Catalog No. 50-295-2430