Learn More
Cayman Chemical ANTIBODY SalubrInal 5mg
SDP
Supplier:  Cayman Chemical 147355 MG
Encompass_Preferred
A selective phosphatase inhibitor which prevents dephosphorylation of eIF2α, protecting cells from ER stress-mediated apoptosis (EC50 = 15 μM); prevents replication of herpes virus (IC50 = 3 μM) and alters bone formation
Catalog No. 50-297-0922