Learn More
Cayman Chemical ANTIBODY SB 204990 5mg
SDP
Supplier:  Cayman Chemical 152455 MG
Encompass_Preferred
A cell-permeable prodrug of SB 201076; inhibits fatty acid and cholesterol synthesis in HepG2 cells in a concentration-dependent manner; reduces plasma levels of the lipoproteins VLDL, LDL, and HDL by 21, 40, and 22%, respectively, in dogs when administered orally at a dose of 25 mg/kg; reduces VLDL synthesis and plasma cholesterol and triglyceride levels in rats in a dose-dependent manner
Catalog No. 50-296-9431