Learn More
Cayman Chemical ANTIBODY SB 225002 5mg
SDP
Supplier:  Cayman Chemical 133365 MG
Encompass_Preferred
A selective non-peptide inhibitor of CXCR2, inhibiting IL-8 binding to CXCR2 (IC50 = 22 nM)
Catalog No. 50-295-1905