Learn More
Cayman Chemical ANTIBODY SB-3CT 1mg
SDP
Supplier:  Cayman Chemical 163371 MG
Encompass_Preferred
A selective, mechanism-based inhibitor of MMP-2 and MMP-9 (Kis = 28 and 400 nM, respectively); a monohydroxylated metabolite has greater inhibitory activity (Kis = 6 and 160 nM for MMP-2 and MMP-9, respectively); can cross the blood-brain barrier
Catalog No. 50-297-0985