Learn More
Cayman Chemical ANTIBODY SCH 28080 50mg
SDP
Supplier:  Cayman Chemical 1788550 MG
Encompass_Preferred
A K+-competitive inhibitor of H+/K+-ATPases that is best known for its ability to block acid secretion through its action against the gastric H+/K+-ATPase (IC50 = 1.3 µM); effective against gastric H+/K+-ATPases from a variety of species and can inhibit some colonic H+/K+-ATPases
Catalog No. 50-298-0348