Learn More
Cayman Chemical ANTIBODY SCH 900776 10mg
SDP
Supplier:  Cayman Chemical 1813110 MG
Encompass_Preferred
A functionally selective inhibitor of Chk1 (Kd = 2 nM; IC50 = 3 nM); interacts synergistically with DNA antimetabolite agents in vitro and in vivo to selectively induce double-strand DNA breaks and cell death in tumor cells
Catalog No. 50-296-6724