Learn More
Cayman Chemical ANTIBODY SenxIn A 25mg
SDP
Supplier:  Cayman Chemical 3029125 MG
Encompass_Preferred
A Cdk8 inhibitor (IC50 = 0.28 µM); inhibits p21WAF1-induced reporter expression in reporter assay using HT-1080 p21-9 cells (IC50 = 0.68 µM); prevents 17β-estradiol-induced increases in expression of mRNA encoding GREB1, CXCL12, and TFF1 in estrogen-deprived MCF-7, BT474, and T47D cancer cells; decreases intracellular and extracellular viral RNA, the number of infectious particles, as well as dengue virus-induced increases in the levels of mRNA encoding LC3-II in dengue virus-infected Huh7 cells at 25 µM; reduces mitochondrial OCR and increases glycolysis in uninfected and dengue virus-infected Huh7 cells at 12.5 µM
Catalog No. 50-298-8879