Learn More
Cayman Chemical ANTIBODY SenxIn B 1mg
SDP
Supplier:  Cayman Chemical 284591 MG
Encompass_Preferred
An inhibitor of Cdk8 and Cdk19; inhibits Cdk8 (IC50s = 24-50 nM, depending on assay); selectively inhibits Cdk19 and Cdk8 by 98.6 and 97.8%, respectively, in a panel of >450 kinases at 2 μM but does inhibit MAP4K2 and YSK4 by 69 and 59%, respectively; inhibits cell growth in MCF-7, BT474, and T47D-ER/Luc breast cancer cells in estrogen-containing media at 1.25-5 μM; reduces tumor growth in an MCF-7 mouse xenograft model at 100 mg/kg twice per day; inhibits RANKL-induced differentiation of murine BMDMs into osteoclasts at 1 and 1.5 μM; increases the bone volume fraction and bone mineral density in injured tibiae in a rat model of cancellous bone injury and regeneration when administered locally at 1 μg
Catalog No. 50-294-1464