Learn More
Cayman Chemical ANTIBODY SGC-CBP30 50mg
SDP
Supplier:  Cayman Chemical 1446950 MG
Encompass_Preferred
A potent inhibitor of CREBBP and EP300 bromodomains (IC50s = 21-69 and 38 nM, respectively)
Catalog No. 50-297-0958