Learn More
Cayman Chemical ANTIBODY SGI-1027 1mg
SDP
Supplier:  Cayman Chemical 111651 MG
Encompass_Preferred
An inhibitor of the DNA methyltransferases DNMT1, DNMT3A, and DNMT3B (IC50s = 12.5, 8, and 7.5 μM, respectively, with Poly(dI-dC) as the substrate); reactivates silenced tumor suppressor genes by reducing CpG island hypermethylation
Catalog No. 50-295-4572