Learn More
Cayman Chemical ANTIBODY SGI-7079 10mg
SDP
Supplier:  Cayman Chemical 2011110 MG
Encompass_Preferred
An Axl kinase inhibitor; inhibits proliferation of inflammatory breast cancer cells (IC50s = 0.43 and 0.15 µM for SUM149 and KPL-4 cells, respectively); synergistically increases the potency of erlotinib on EGFR inhibition; dose-dependently inhibits tumor growth in a mouse xenograft model of non-small cell lung cancer
Catalog No. 50-298-0795