Learn More
Cayman Chemical ANTIBODY SMI-16a 10mg
SDP
Supplier:  Cayman Chemical 2945610 MG
Encompass_Preferred
A Pim-1 kinase inhibitor (IC50 = 63 nM); selective for Pim-1 over a panel of 60 kinases; inhibits phosphorylation of the Pim-1 target protein Bad in DU145-Pim cells and inhibits the growth of PC3, DU145, LNCaP, K562, and MV4-11 cancer cells at 5 μM
Catalog No. 50-299-2384