Learn More
Cayman Chemical ANTIBODY SMI-481 5mg
SDP
Supplier:  Cayman Chemical 176375 MG
Encompass_Preferred
An inhibitor of the yeast PITP Sec14, blocking both Sec14-mediated transfer of [3H]-PtdIns in vitro and growth (IC50s = 0.21 and 2.87 µM, respectively)
Catalog No. 50-298-0086