Learn More
Cayman Chemical ANTIBODY SnT 1 10mg
SDP
Supplier:  Cayman Chemical 1493310 MG
Encompass_Preferred
A potent inhibitor of hedgehog signaling that acts by directly antagonizing SMO (IC50 = 20 nM); counteracts SMO activation by the SMO agonist SAG but only partially competes with SAG binding to SMO
Catalog No. 50-297-2908