Learn More
Cayman Chemical ANTIBODY SpautIn-1 5mg
SDP
Supplier:  Cayman Chemical 177695 MG
Encompass_Preferred
An autophagy inhibitor, inducing cell death in human breast cancer Bcap-37 cells grown in glucose-free media (EC50 = ~1.25 µM); promotes the degradation of Vps34 PI3K complex by inhibiting the deubiquitinating enzymes USP10 and USP13 (IC50s = 0.58 and 0.69 µM, respectively)
Catalog No. 50-296-6717