Learn More
Cayman Chemical ANTIBODY SPI 112 25mg
SDP
Supplier:  Cayman Chemical 2947425 MG
Encompass_Preferred
A cell-impermeable inhibitor of SHP-2 in cell-free assays (IC50 = 1 µM); selectively inhibits SHP-2 over SHP-1 and PTP1B (IC50s = 18.3 and 14.5 µM, respectively)
Catalog No. 50-299-1239