Learn More
Cayman Chemical ANTIBODY SR 0987 5mg
SDP
Supplier:  Cayman Chemical 195035 MG
Encompass_Preferred
An agonist of RORγt, inducing reporter gene expression with an EC50 value of ~800 nM; increases expression of IL-17, while decreasing expression of PD-1, in a variety of T cells
Catalog No. 50-298-2229