Learn More
Cayman Chemical ANTIBODY SR 1078 1mg
SDP
Supplier:  Cayman Chemical 165031 MG
Encompass_Preferred
A selective RORα and RORγ agonist that stimulates ROR transcriptional activity in HEK293 cell reporter assays at concentrations as low as 2 µM without effect at the related LXR and FXR receptors; stabilizes p53 and induces apoptosis in HepG2 cancer cells
Catalog No. 50-296-7463