Learn More
Cayman Chemical ANTIBODY SR-12813 50mg
SDP
Supplier:  Cayman Chemical 1811550 MG
Encompass_Preferred
A hypocholesterolemic compound that promotes the degradation of HMG-CoA reductase in various animal models; human and rabbit pregnane X receptor agonist (Kd = 41 nM; EC50 = 137 nM for hPXR in vitro) that can induce CYP3A expression in human and rabbit hepatocytes
Catalog No. 50-297-9709