Learn More
Cayman Chemical ANTIBODY SR 16832 5mg
SDP
Supplier:  Cayman Chemical 276325 MG
Encompass_Preferred
A dual-site covalent antagonist of PPARγ; inhibits MRL20-induced allosteric activation of PPARγ in a reporter assay using HEK293T cells at 5 μM; reduces basal activity of PPARγ and inhibits binding of DHA to PPARγ in a TR-FRET assay
Catalog No. 50-293-7725