Learn More
Cayman Chemical ANTIBODY SU 4942 50mg
SDP
Supplier:  Cayman Chemical 3296750 MG
Encompass_Preferred
A modulator of tyrosine kinase signaling; inhibits VEGF- or acidic FGF-induced mitogenesis of endothelial cells in vitro
Catalog No. 50-295-0938