Learn More
Cayman Chemical ANTIBODY SU11274 500ug
SDP
Supplier:  Cayman Chemical 14861500 UG
Encompass_Preferred
A potent, selective, ATP-competitive inhibitor of c-Met (IC50 = 20 nM); induces apoptosis and cell cycle arrest in transformed Ba/F3 cells and cancer cell lines
Catalog No. 50-296-8719