Learn More
Cayman Chemical ANTIBODY Suprofen 25mg
SDP
Supplier:  Cayman Chemical 2135025 MG
Encompass_Preferred
An NSAID that inhibits both isoforms of COX (IC50s = 1.1 and 8.7 µM for COX-1 and COX-2, respectively)
Catalog No. 50-298-1488