Learn More
Cayman Chemical ANTIBODY SW033291 25mg
SDP
Supplier:  Cayman Chemical 1804025 MG
Encompass_Preferred
A potent inhibitor of 15-PGDH (Ki = ~0.1 nM) that has activity in vivo; rapidly increases the levels of PGE2 in bone marrow and other tissues in mice after intraperitoneal injection
Catalog No. 50-297-9739