Learn More
Cayman Chemical ANTIBODY T-5342126 10mg
SDP
Supplier:  Cayman Chemical 3309910 MG
Encompass_Preferred
A TLR4 antagonist; reduces LPS-induced NO production in RAW 264.7 cells (IC50 = 27.8 µM) and LPS-induced IL-8, TNF-α, and IL-6 production in isolated human whole blood (IC50s = 110.5, 315.6, and 318.4 µM, respectively); reduces ethanol intake and the abundance of Iba1 in the central nucleus of the amygdala in ethanol-dependent mice at 82 mg/kg
Catalog No. 50-295-0201