Learn More
Cayman Chemical ANTIBODY T-705 100mg
SDP
Supplier:  Cayman Chemical 23384100 MG
Encompass_Preferred
An antiviral agent; inhibits influenza A, B, and C (IC50s = 0.013-0.48 UG/ml); inhibits a GS4071-resistant influenza strain (IC50 = 0.095 UG/ml) and RNA viruses (IC50s = 4.8-41 UG/ml); not active against DNA viruses; undergoes phosphoribosylation to form T-705-RTP, the active form; decreases the rate of mortality in influenza-infected mice when used at a dose of 200 mg/kg once daily; reduces the lung virus yield in a dose-dependent manner at doses ranging from 50-200 mg/kg per day in influenza-infected mice
Catalog No. 50-299-3731