Learn More
Cayman Chemical ANTIBODY T-863 1mg
SDP
Supplier:  Cayman Chemical 258071 MG
Encompass_Preferred
A DGAT-1 inhibitor (IC50 = 15 nM); selective for DGAT-1 over DGAT-2 and ACAT-1 (IC50s = >10 and 12 μM, respectively); inhibits triacylglycerol synthesis in HuTu 80 cells (IC50 = 4 nM); reduces plasma triacylglycerol levels, the ratio of triacylglycerols:diacylglycerols, and body weight in a mouse model of diet-induced obesity at 20 mg/kg
Catalog No. 50-293-6982