Learn More
Cayman Chemical ANTIBODY TA-01 5mg
SDP
Supplier:  Cayman Chemical 246675 MG
Encompass_Preferred
An inducer of cardiomyocyte differentiation; increases expression of the cardiomyocyte marker NKX2-5 by 2.2-fold and decreases expression of mesoderm markers and the pre-cardiac marker Isl-1 in HES-3 NKX2-5eGFP/w cells at <5 μM; inhibits CK1ε and CK1Δ and reduces expression of Wnt/β-catenin signaling pathway members
Catalog No. 50-293-6297