Learn More
Cayman Chemical ANTIBODY TafamIdIs 25mg
SDP
Supplier:  Cayman Chemical 1369425 MG
Encompass_Preferred
A TTR kinetic stabilizer; binds to TTR with negative cooperativity (Kd1 = 3 nM; Kd2 = 278 nM) to stabilize the TTR dimer-dimer interface and inhibit tetrameric dissociation; stabilizes wild-type and clinically significant V30M and V122I mutant TTR amyloidogenic homotetramers (EC50s = 2.7-3.2 μM) under fibril-promoting, denaturing, and physiological conditions in vitro; stabilizes TTR heterotetramers containing wild-type and mutant subunits ex vivo in human plasma derived from patients carrying V30M or V1221 mutations when used at a concentration of 7.2 μM
Catalog No. 50-295-5107