Learn More
Cayman Chemical ANTIBODY TarIquIdar 10mg
SDP
Supplier:  Cayman Chemical 2418010 MG
Encompass_Preferred
A P-gp inhibitor; binds to P-glycoprotein (Kd = 5.1 nM); inhibits transport of vinblastine and paclitaxel in CHrB30 cells, leading to accumulation to control levels (EC50 = 487 nM); increases the amount of its substrate that enters the CNS
Catalog No. 50-293-4258