Learn More
Cayman Chemical ANTIBODY TAS 0728 25mg
SDP
Supplier:  Cayman Chemical 3352825 MG
Encompass_Preferred
A covalent HER2 inhibitor (IC50 = 36 nM); selective for HER2 over panels of 386 and 374 additional kinases, as well as a panel of 68 non-kinase enzymes, at 1 µM; inhibits HER2 autophosphorylation in HER2-overexpressing SK-BR-3 cells; inhibits the growth of six HER2-amplified cancer cell lines (GI50s = 1.6-31 nM); reduces tumor volume in NCI N87 and BT474 mouse xenograft models
Catalog No. 50-294-9504